Learn More
TARGETMOL CHEMICALS INC NORATHYRIOL 5MG
SDP
Catalog No. 502578886
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Norathyriol is a potent PTP1B inhibitor with good cell permeability and oral availability. purity: 98%
Catalog No. 50-257-8886 Supplier TARGETMOL CHEMICALS INC Supplier No. TN19905MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Norathyriol is a potent PTP1B inhibitor with good cell permeability and oral availability. purity: 98%