Learn More
Enterprise Technology Solutions Norazza Inc Electronics Cleaner,Spray,Multisurface,Anti-Static,8oz,BE
SDP
Catalog No. 5000740708
Encompass_Preferred
Anti-Static Multisurface Electronics Cleaner is specially formulated to safely and effectively clean all types of screens and all external electronic surfaces. Spray removes dust and gently cleans away fingerprints. It even prevents buildup of static charges. Formula is free of oil, wax and ammonia. Spray doesn't run when it's applied so you can clean where you spray it. It cleans your valuable electronics including tablets, TVs, computers, phones and more and leaves a fresh scent. Convenient size fits easily in your desk drawer.
Catalog No. 50-007-40708 Supplier Enterprise Technology Solutions Supplier No. NRZ097000
Anti-Static Multisurface Electronics Cleaner is specially formulated to safely and effectively clean all types of screens and all external electronic surfaces. Spray removes dust and gently cleans away fingerprints. It even prevents buildup of static charges. Formula is free of oil, wax and ammonia. Spray doesn't run when it's applied so you can clean where you spray it. It cleans your valuable electronics including tablets, TVs, computers, phones and more and leaves a fresh scent. Convenient size fits easily in your desk drawer.