Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Norbergenin | 79595-97-4 | MFCD09970989 | 98.3% | 314.24 | 10 MG
SDP

Catalog No. 5000425783
Encompass_Preferred

Norbergenin, an O-demethyl derivative of bergenin, exhibits moderate antioxidant activity. It shows an IC50 of 13 μM in DPPH radical scavenging and 32 μM in superoxide anion scavenging.

  • Purity: 98.31%
  • Molecular weight: 314.24
  • Chemical formula: C13H14O9
  • CAS number: 79595-97-4
  • Appears as a solid, white to yellow in color
  • Soluble in DMSO at 125 mg/mL
  • Soluble in various aqueous solutions, including 10% DMSO, 40% PEG300, 5% Tween-80, 45% Saline (≥ 2.08 mg/mL)
  • Also soluble in 10% DMSO, 90% (20% SBE-β-CD in Saline) (≥ 2.08 mg/mL)
  • And in 10% DMSO, 90% Corn Oil (≥ 2.08 mg/mL)

Catalog No. 50-004-25783 Supplier Medchemexpress LLC Supplier No. HYN944710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Norbergenin, an O-demethyl derivative of bergenin, exhibits moderate antioxidant activity. It shows an IC50 of 13 μM in DPPH radical scavenging and 32 μM in superoxide anion scavenging.

  • Purity: 98.31%
  • Molecular weight: 314.24
  • Chemical formula: C13H14O9
  • CAS number: 79595-97-4
  • Appears as a solid, white to yellow in color
  • Soluble in DMSO at 125 mg/mL
  • Soluble in various aqueous solutions, including 10% DMSO, 40% PEG300, 5% Tween-80, 45% Saline (≥ 2.08 mg/mL)
  • Also soluble in 10% DMSO, 90% (20% SBE-β-CD in Saline) (≥ 2.08 mg/mL)
  • And in 10% DMSO, 90% Corn Oil (≥ 2.08 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.