Learn More
Medchemexpress LLC HY-100903 10mg , Norbinaltorphimine dihydrochloride CAS:113158-34-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10090310MG
Encompass_Preferred
HY-100903 10mg Norbinaltorphimine dihydrochloride CAS: 113158-34-2 Norbinaltorphimine dihydrochloride is a potent and selective κ opioid receptor antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9874