Learn More
Medchemexpress LLC Norbinaltorphimine dihydrochloride | 113158-34-2 | MFCD00069374 | >98.0% | 734.71 g/mol | C40H45Cl2N3O6 | 1 ML
SDP
Catalog No. 5000212327
Encompass_Preferred
Norbinaltorphimine dihydrochloride is a selective κ-opioid receptor antagonist used in pharmacology research to probe kappa opioid receptor function. The dihydrochloride salt has CAS 113158-34-2 and a molecular weight of 734.71 g/mol; it is typically supplied at high purity for in vitro studies and is available as a ready-to-use 10 mM solution in DMSO.
  • Selective kappa-opioid receptor antagonist for receptor pharmacology studies.
  • Supplied as a dihydrochloride salt for improved solubility.
  • High purity suitable for in vitro assays.
  • Available as ready-to-use solution (10 mM) to simplify dosing.
  • Store sealed, protect from moisture, and keep solution aliquots frozen.
Catalog No. 50-002-12327 Supplier Medchemexpress LLC Supplier No. HY10090310MM 1ML
Norbinaltorphimine dihydrochloride is a selective κ-opioid receptor antagonist used in pharmacology research to probe kappa opioid receptor function. The dihydrochloride salt has CAS 113158-34-2 and a molecular weight of 734.71 g/mol; it is typically supplied at high purity for in vitro studies and is available as a ready-to-use 10 mM solution in DMSO.
  • Selective kappa-opioid receptor antagonist for receptor pharmacology studies.
  • Supplied as a dihydrochloride salt for improved solubility.
  • High purity suitable for in vitro assays.
  • Available as ready-to-use solution (10 mM) to simplify dosing.
  • Store sealed, protect from moisture, and keep solution aliquots frozen.