Learn More
Cayman Chemical Norbixin (hydrate), Purity: ≥70%, 5 mg
SDP
Catalog No. 502944899
Encompass_Preferred
A carotenoid with diverse biological activities; binds to PPARγ in a cell-free assay (Ki = 1.15 µM); attenuates hyperglycemia, hyperinsulinemia, and insulin resistance, as well as decreases serum lipid levels and cardiac levels of TBARS and GSH in a rat model of cardio-metabolic syndrome at 47.7 mg/kg; decreases serum levels of oxidized LDL and aortic protein oxidation, as well as reduces atherosclerotic area, in a rabbit model of cholesterol-induced atherosclerosis; reduces mercury-induced DNA damage in isolated rat hepatocytes and leukocytes at 0.1 and 1 mg/kg; prevents photoreceptor degeneration in an Abca4-/- Rdh8-/- mouse model of AMD
Catalog No. 50-294-4899 Supplier Cayman Chemical Supplier No. 401325 MG
A carotenoid with diverse biological activities; binds to PPARγ in a cell-free assay (Ki = 1.15 µM); attenuates hyperglycemia, hyperinsulinemia, and insulin resistance, as well as decreases serum lipid levels and cardiac levels of TBARS and GSH in a rat model of cardio-metabolic syndrome at 47.7 mg/kg; decreases serum levels of oxidized LDL and aortic protein oxidation, as well as reduces atherosclerotic area, in a rabbit model of cholesterol-induced atherosclerosis; reduces mercury-induced DNA damage in isolated rat hepatocytes and leukocytes at 0.1 and 1 mg/kg; prevents photoreceptor degeneration in an Abca4-/- Rdh8-/- mouse model of AMD