Learn More
Medchemexpress LLC Norbornene | 498-66-8 | 99.9% | 94.16 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW013021100G
Encompass_Preferred
Norbornene is a bicyclic compound. Norbornene polymers can be used in tissue engineering and drug release systems.
    Catalog No. 50-004-51393