Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Norbornene | 498-66-8 | MFCD00082304 | 100.0% | 94.16 | 500 G
SDP

Catalog No. 5000451997
Encompass_Preferred

Norbornene is a bicyclic compound whose polymers can be used in tissue engineering and drug release systems.

  • Purity of 99.95%
  • Molecular weight is 94.16
  • Chemical formula is C7H10
  • CAS number is 498-66-8
  • Appears as a solid below 44°C and liquid above 46°C
  • Color is colorless to off-white
  • Solubility in DMSO is 100 mg/mL (1062.02 mM)
  • Store pure form at -20°C for 3 years or 4°C for 2 years

Catalog No. 50-004-51997 Supplier Medchemexpress LLC Supplier No. HYW013021500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Norbornene is a bicyclic compound whose polymers can be used in tissue engineering and drug release systems.

  • Purity of 99.95%
  • Molecular weight is 94.16
  • Chemical formula is C7H10
  • CAS number is 498-66-8
  • Appears as a solid below 44°C and liquid above 46°C
  • Color is colorless to off-white
  • Solubility in DMSO is 100 mg/mL (1062.02 mM)
  • Store pure form at -20°C for 3 years or 4°C for 2 years

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.