Learn More
Medchemexpress LLC Norbornene-NHS | 1888335-48-5 | C12H13NO5 | 100 MG
SDP
Catalog No. 5000374205
Encompass_Preferred
Norbornene-NHS is a click chemistry reagent that functions as a dienophile. It is utilized for conjugation through copper-free Click reactions with tetrazines.
  • Classified as a click chemistry reagent.
  • Acts as a dienophile.
  • Used for conjugation.
  • Conjugation occurs via copper-free click reactions.
  • Reacts with tetrazines in the conjugation process.
  • For research use only and not intended for human use.
Catalog No. 50-003-74205 Supplier Medchemexpress LLC Supplier No. HY151727100MG
Norbornene-NHS is a click chemistry reagent that functions as a dienophile. It is utilized for conjugation through copper-free Click reactions with tetrazines.
  • Classified as a click chemistry reagent.
  • Acts as a dienophile.
  • Used for conjugation.
  • Conjugation occurs via copper-free click reactions.
  • Reacts with tetrazines in the conjugation process.
  • For research use only and not intended for human use.