Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Norbornene-NHS | 1888335-48-5 | C12H13NO5 | 50 MG
SDP

Catalog No. 5000374125
Encompass_Preferred

Norbornene-NHS is a click chemistry reagent and a dienophile for conjugation via copper-free Click reaction with tetrazines. It is for research use only and not sold to patients.

  • Purity: 92.08%
  • Molecular weight: 251.24
  • Appearance: Liquid
  • Color: Light yellow to yellow
  • Used for conjugation via copper-free click reaction

Catalog No. 50-003-74125 Supplier Medchemexpress LLC Supplier No. HY15172750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Norbornene-NHS is a click chemistry reagent and a dienophile for conjugation via copper-free Click reaction with tetrazines. It is for research use only and not sold to patients.

  • Purity: 92.08%
  • Molecular weight: 251.24
  • Appearance: Liquid
  • Color: Light yellow to yellow
  • Used for conjugation via copper-free click reaction

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.