Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Norbornylene, 99%, stabilized, ACROS Organics™

Click to view available options
Quantity:
25g
500g
Packaging:
Glass bottle
498-66-8
C7H10
94.15
MFCD00082304
JFNLZVQOOSMTJK-UHFFFAOYSA-N
bicyclo 2.2.1 hept-2-ene,norbornylene,2-norbornene,norbornene,norcamphene,norfenchene,2-norbornylene,3,6-endomethylenecyclohexene,bicyclo 2.2.1-2-heptene,bicyclo 2.2.1.-2-heptene
10352
bicyclo[2.2.1]hept-2-ene
C1CC2CC1C=C2
498-66-8
C7H10
94.15
MFCD00082304
JFNLZVQOOSMTJK-UHFFFAOYSA-N
bicyclo 2.2.1 hept-2-ene,norbornylene,2-norbornene,norbornene,norcamphene,norfenchene,2-norbornylene,3,6-endomethylenecyclohexene,bicyclo 2.2.1-2-heptene,bicyclo 2.2.1.-2-heptene
10352
bicyclo[2.2.1]hept-2-ene
C1CC2CC1C=C2
Packaging Glass bottle
Quantity 25g
Flash Point −15°C
Color White
Boiling Point 96.0°C
Stabilizer 0.1% phenolic antioxidant
Melting Point 42.0°C to 46.0°C
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Fieser 01,757; 02,300
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in toluene and benzene
Formula Weight 94.15
Physical Form Crystalline Mass
Percent Purity 99%
Chemical Name or Material Norbornylene, 99%
Show More Show Less

GHS P Statement
Flammable solid.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 207-866-0

RTECSNumber : RB7900000

TSCA : TSCA