Learn More
Selleck Chemical LLC Norcantharidin S3759-25mg
SDP
Catalog No. 503109749
Encompass_Preferred
Norcantharidin (Endothall anhydride) is a synthetic anticancer compound which is a dual inhibitor for c-Met and EGFR in human colon cancers
Catalog No. 50-310-9749 Supplier Selleck Chemical LLC Supplier No. S375925MG
Norcantharidin (Endothall anhydride) is a synthetic anticancer compound which is a dual inhibitor for c-Met and EGFR in human colon cancers