Learn More
TARGETMOL CHEMICALS INC Norcantharidin 50MG
SDP
Catalog No. 502877005
Encompass_Preferred
Also available in 1 g, 5 mg, 10 mg, 25 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. (±)-Norcantharidin (Endothall anhydride) is a synthetic anticancer compound which is a dual inhibitor for c-Met and EGFR in human colon cancers. Purity ≥98%
Catalog No. 50-287-7005 Supplier TARGETMOL CHEMICALS INC Supplier No. T565650MG
Also available in 1 g, 5 mg, 10 mg, 25 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. (±)-Norcantharidin (Endothall anhydride) is a synthetic anticancer compound which is a dual inhibitor for c-Met and EGFR in human colon cancers. Purity ≥98%