Learn More
Cayman Chemical Norcholic Acid, Purity: ≥95%, CAS: 60696-62-0, 100 mg
SDP
Catalog No. 502947756
Encompass_Preferred
A bile acid and derivative of cholic acid; urine levels are increased in patients with liver cirrhosis or CTX
Catalog No. 50-294-7756 Supplier Cayman Chemical Supplier No. 34623100 MG
A bile acid and derivative of cholic acid; urine levels are increased in patients with liver cirrhosis or CTX