Learn More
Cayman Chemical Norcantharidin, Purity: ≥95%, CAS: 29745-04-8, 1 g
SDP
Catalog No. 502939150
Encompass_Preferred
A PP1 and PP2A inhibitor (IC50s = 9 and 3 µM, respectively); induces cell cycle arrest at the G2/M phase and apoptosis in HepG2, Hep3B, and Huh7 human hepatoma cell lines at 5 µg/ml; reduces angiogenesis in a chick chorioallantoic membrane assay at 500-5,000 µg/ml; inhibits growth in a panel of nine human cancer cell lines (GI50s = 30-64 µM); reduces tumor growth in a GBC-SD mouse xenograft model at 28 mg/kg
Catalog No. 50-293-9150 Supplier Cayman Chemical Supplier No. 284571 G
A PP1 and PP2A inhibitor (IC50s = 9 and 3 µM, respectively); induces cell cycle arrest at the G2/M phase and apoptosis in HepG2, Hep3B, and Huh7 human hepatoma cell lines at 5 µg/ml; reduces angiogenesis in a chick chorioallantoic membrane assay at 500-5,000 µg/ml; inhibits growth in a panel of nine human cancer cell lines (GI50s = 30-64 µM); reduces tumor growth in a GBC-SD mouse xenograft model at 28 mg/kg