Learn More
Medchemexpress LLC Norcyclizine (1-(diphenylmethyl)piperazine) | 841-77-0 | 99.7% | 252.35 | 50 G
SDP
Catalog No. 5000454753
Encompass_Preferred
Norcyclizine is a piperazine compound utilized in the synthesis of antimicrobial agents. Derivatives of 1-Benzhydrylpiperazine have demonstrated an ability to enhance the antibacterial activity of β-lactam antibiotics, such as Oxacillin, against Methicillin-resistant Staphylococcus aureus (MRSA). This enhancement is believed to occur through the inhibition of the allosteric site of PBP2a. Furthermore, 1-Benzhydrylpiperazine can also function as a pharmacological scaffold for the development of anticancer agents.
  • Used for the synthesis of antimicrobial agents.
  • Derivatives enhance antibacterial activity of β-lactam antibiotics against MRSA.
  • Likely inhibits the allosteric site of PBP2a.
  • Can serve as a pharmacological scaffold for anticancer agents.
Catalog No. 50-004-54753 Supplier Medchemexpress LLC Supplier No. HYW01337650G
Add to Cart
Add to Cart
Norcyclizine is a piperazine compound utilized in the synthesis of antimicrobial agents. Derivatives of 1-Benzhydrylpiperazine have demonstrated an ability to enhance the antibacterial activity of β-lactam antibiotics, such as Oxacillin, against Methicillin-resistant Staphylococcus aureus (MRSA). This enhancement is believed to occur through the inhibition of the allosteric site of PBP2a. Furthermore, 1-Benzhydrylpiperazine can also function as a pharmacological scaffold for the development of anticancer agents.
  • Used for the synthesis of antimicrobial agents.
  • Derivatives enhance antibacterial activity of β-lactam antibiotics against MRSA.
  • Likely inhibits the allosteric site of PBP2a.
  • Can serve as a pharmacological scaffold for anticancer agents.