Learn More
Medchemexpress LLC Norcyclizine (1-(Diphenylmethyl)piperazine) | 841-77-0 | 99.7% | 252.35 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW013376500G
Encompass_Preferred
Norcyclizine is a piperazine compound utilized in the synthesis of antimicrobial and anticancer agents. Its derivatives, specifically 1-Benzhydrylpiperazine, are known to enhance the antibacterial activity of β-lactam antibiotics against Methicillin-resistant Staphylococcus aureus (MRSA) by inhibiting the allosteric site of PBP2a. This compound also serves as a pharmacological scaffold for developing anticancer agents. This product is intended for research use only.
  • Used for synthesizing antimicrobial agents.
  • Enhances antibacterial activity of β-lactam antibiotics against MRSA.
  • Inhibits the allosteric site of PBP2a.
  • Serves as a pharmacological scaffold for synthesizing anticancer agents.
  • Intended for research use only.
Catalog No. 50-004-54294