Learn More
Medchemexpress LLC Norcyclizine | 841-77-0 | 99.7% | 252.35 | 25 G
SDP
Catalog No. 5000366756
Encompass_Preferred
Norcyclizine is a piperazine compound used for the synthesis of antimicrobial and anticancer agents. It enhances the antibacterial activity of β-lactam antibiotics against Methicillin-resistant Staphylococcus aureus (MRSA) by inhibiting the allosteric site of PBP2a.
  • Used for the synthesis of antimicrobial agents
  • Enhances antibacterial activity of β-lactam antibiotics against MRSA
  • Inhibits the allosteric site of PBP2a
  • Serves as a pharmacological scaffold for synthesizing anticancer agents
Catalog No. 50-003-66756 Supplier Medchemexpress LLC Supplier No. 25GHYW013376
Norcyclizine is a piperazine compound used for the synthesis of antimicrobial and anticancer agents. It enhances the antibacterial activity of β-lactam antibiotics against Methicillin-resistant Staphylococcus aureus (MRSA) by inhibiting the allosteric site of PBP2a.
  • Used for the synthesis of antimicrobial agents
  • Enhances antibacterial activity of β-lactam antibiotics against MRSA
  • Inhibits the allosteric site of PBP2a
  • Serves as a pharmacological scaffold for synthesizing anticancer agents