Learn More
Cayman Chemical NordeoxycholIc AcId 100mg
SDP
Catalog No. 502989649
Encompass_Preferred
A metabolite of norcholic acid and derivative of deoxycholic acid; hepatic levels are decreased in rats in a high-fat diet model of NAFLD; serum levels are lower in men compared with women; has commonly been used as an internal standard for the quantification of bile acids in various sample types by GC- and LC-MS
Catalog No. 50-298-9649 Supplier Cayman Chemical Supplier No. 30837100 MG
A metabolite of norcholic acid and derivative of deoxycholic acid; hepatic levels are decreased in rats in a high-fat diet model of NAFLD; serum levels are lower in men compared with women; has commonly been used as an internal standard for the quantification of bile acids in various sample types by GC- and LC-MS