Learn More
Cayman Chemical NordeoxycholIc AcId 10mg
SDP
Catalog No. 502990393
Encompass_Preferred
A metabolite of norcholic acid and derivative of deoxycholic acid; hepatic levels are decreased in rats in a high-fat diet model of NAFLD; serum levels are lower in men compared with women; has commonly been used as an internal standard for the quantification of bile acids in various sample types by GC- and LC-MS
Catalog No. 50-299-0393 Supplier Cayman Chemical Supplier No. 3083710 MG
A metabolite of norcholic acid and derivative of deoxycholic acid; hepatic levels are decreased in rats in a high-fat diet model of NAFLD; serum levels are lower in men compared with women; has commonly been used as an internal standard for the quantification of bile acids in various sample types by GC- and LC-MS