Learn More
Cayman Chemical Nordeoxycholic Acid, Purity: ≥95%, CAS: 53608-86-9, 50 mg
SDP
Catalog No. 502989995
Encompass_Preferred
A metabolite of norcholic acid and derivative of deoxycholic acid; hepatic levels are decreased in rats in a high-fat diet model of NAFLD; serum levels are lower in men compared with women; has commonly been used as an internal standard for the quantification of bile acids in various sample types by GC- and LC-MS
Catalog No. 50-298-9995 Supplier Cayman Chemical Supplier No. 3083750 MG
A metabolite of norcholic acid and derivative of deoxycholic acid; hepatic levels are decreased in rats in a high-fat diet model of NAFLD; serum levels are lower in men compared with women; has commonly been used as an internal standard for the quantification of bile acids in various sample types by GC- and LC-MS