Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Radians Nordic Blaze™ 3-in-1 Thermal Balaclava
SDP

Catalog No. 19035294
Change view
Click to view available options
Material:
SE-treated Polyester
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-035-294 SE-treated Polyester
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-035-294 Supplier Radians Supplier No. RWL22SE
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protects the face and neck from cold, harsh elements

  • Made of Nordic thermal fleece for added warmth and comfort
  • Converts from balaclava to neck gator or face shield
  • Erogonomic design to combat wearer fatigue
  • SE-treated polyester
    • Treated to be self-extinguishing as defined by ASTM D6413

Specifications

Product Type 3-in-1 Balaclava
Material SE-treated Polyester
Color Black
For Use With (Application) For warmth and protection from harsh conditions

Warning: Cancer and reproductive harm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.