Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Radians Nordic Blaze™ Thermal Hard Hat Liner
SDP

Catalog No. 19035293
Catalog No. 19-035-293 Supplier Radians Supplier No. RWL19
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protects the head and neck from cold, harsh elements

  • Dual layer liner allows for added warmth
  • Outer layer blocks out wind
  • Inner Nordic fleece thermal layer keeps in heat
  • Lightweight, ergonomic design for added comfort
  • Hook and loop closure easily attaches to internal hard hat system
  • Chin strap for stability while wearing hard hats

Specifications

Product Type Thermal Hard Hat Liner
Color Black, Red
Closure Type Hook and Loop
Includes Chin Strap
For Use With (Application) For keeping heat in and blocking out wind

Warning: Cancer and reproductive harm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.