Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nordihydorguaiaretic acid, 99%, For HPLC analysis, MP Biomedicals™

Catalog No. ICN15594491 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15594491 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15594491 Supplier MP Biomedicals, Inc Supplier No. 0215594491
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 500-38-9
Molecular Formula C18H22O4
Molecular Weight (g/mol) 302.37
InChI Key HCZKYJDFEPMADG-UHFFFAOYSA-N
Synonym nordihydroguaiaretic acid, ndga, dihydronorguaiaretic acid, norhydroguaiaretic acid, nordihydroguairaretic acid, norguaiaretic acid, dihydro, 1,2-benzenediol, 4,4'-2,3-dimethyl-1,4-butanediyl bis, dinorguaiaretic acid, dihydro, 4,4'-2,3-dimethyltetramethylene dipyrocatechol, 2,3-bis 3,4-dihydroxyphenylmethyl butane
PubChem CID 4534
ChEBI CHEBI:7625
IUPAC Name 4-[4-(3,4-dihydroxyphenyl)-2,3-dimethylbutyl]benzene-1,2-diol
SMILES CC(CC1=CC(=C(C=C1)O)O)C(C)CC2=CC(=C(C=C2)O)O

Specifications

Color White
Quantity 1 g
Formula Weight 302.4
Percent Purity 0.99
Physical Form Crystalline Powder
Chemical Name or Material Nordihydorguaiaretic acid, For HPLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.