Learn More
AdipoGen Nordihydrocapsiate
SDP
Supplier:  AdipoGen CDXN0121M100
Encompass_Preferred
Chemical. CAS 220012-53-3. Formula C17H26O4. MW 294.39. Synthetic Major capsaicinoid. TRPV1 and hTRPA1 agonist. Shown to activate capsaicin receptor, also called transient receptor potential vanilloid subfamily member 1 TRPV1, but without inducing a nociceptive response. Anti-inflammatory and anticancer compound.
Catalog No. 50-256-0405