Learn More
Cayman Chemical Nordihydroguaiaretic Acid, Purity: ≥95%, CAS: 500-38-9, 500 mg
SDP
Catalog No. 502893505
Encompass_Preferred
NDGA is a non-selective lipoxygenase inhibitor which blocks CysLT synthesis. NDGA inhibits A-23187-induced CysLT biosynthesis in rat peritoneal cells with an IC50 value of 5-7 µM.{346} The IC50 values are 3.0-5.0 µM for human platelet 12-LO and 0.91 µM for rabbit reticulocyte 15-LO.{346,1462}
Catalog No. 50-289-3505 Supplier Cayman Chemical Supplier No. 70300500 MG
NDGA is a non-selective lipoxygenase inhibitor which blocks CysLT synthesis. NDGA inhibits A-23187-induced CysLT biosynthesis in rat peritoneal cells with an IC50 value of 5-7 µM.{346} The IC50 values are 3.0-5.0 µM for human platelet 12-LO and 0.91 µM for rabbit reticulocyte 15-LO.{346,1462}