Learn More
Medchemexpress LLC HY-N0198 100mg Medchemexpress, Nordihydroguaiaretic acid CAS:500-38-9 Purity:>98%
SDP
Catalog No. 501933461
Encompass_Preferred
Medchemexpress, HY-N0198 100mg Nordihydroguaiaretic acid CAS:500-38-9 Nordihydroguaiaretic acid is a 5-lipoxygenase ( 5LOX ) ( IC 50 =8±3 μM) and tyrosine kinase inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3461 Supplier Medchemexpress LLC Supplier No. HYN0198100MG
Medchemexpress, HY-N0198 100mg Nordihydroguaiaretic acid CAS:500-38-9 Nordihydroguaiaretic acid is a 5-lipoxygenase ( 5LOX ) ( IC 50 =8±3 μM) and tyrosine kinase inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.