Learn More
Medchemexpress LLC Nordoxepin-d3 hydrochloride | 1331665-54-3 | 99.9% | 304.83 g/mol | C18H17D3ClNO | 5 MG
SDP
Catalog No. 5000262584
Encompass_Preferred
Nordoxepin-d3 hydrochloride is the deuterium-labeled hydrochloride salt of nordoxepin (N-desmethyldoxepin). It is provided as a white to off-white solid, high-purity reference standard intended for analytical chemistry, mass spectrometry, and pharmacokinetic or metabolic studies.
  • High isotopic labeling for reliable mass spectrometric internal standard
  • High purity (≈99.9%) for accurate quantitative analysis
  • Hydrochloride salt with solid, white to off-white appearance
  • Suitable for pharmacokinetic and metabolic studies
  • Available in small milligram packaging for standards preparation
Catalog No. 50-002-62584 Supplier Medchemexpress LLC Supplier No. HY133771AS5MG
Nordoxepin-d3 hydrochloride is the deuterium-labeled hydrochloride salt of nordoxepin (N-desmethyldoxepin). It is provided as a white to off-white solid, high-purity reference standard intended for analytical chemistry, mass spectrometry, and pharmacokinetic or metabolic studies.
  • High isotopic labeling for reliable mass spectrometric internal standard
  • High purity (≈99.9%) for accurate quantitative analysis
  • Hydrochloride salt with solid, white to off-white appearance
  • Suitable for pharmacokinetic and metabolic studies
  • Available in small milligram packaging for standards preparation