Learn More
Medchemexpress LLC Nordoxepin-d3 hydrochloride | 1331665-54-3 | 99.9% | C18H17D3ClNO | 10MG
SDP
Catalog No. 5000258204
Encompass_Preferred
Nordoxepin-d3 hydrochloride is the deuterium-labeled form of nordoxepin hydrochloride, the active metabolite of doxepin. It is provided as a solid for use as a tracer and internal standard in quantitative analytical workflows, including NMR, GC-MS, and LC-MS.
  • High chemical purity (99.9%).
  • Deuterium-labeled for accurate internal standardization.
  • Suitable for NMR, GC-MS, and LC-MS quantitation.
  • Supplied as a stable solid for convenient handling and storage.
  • Useful as a tracer in pharmacokinetic and metabolite studies.
Catalog No. 50-002-58204 Supplier Medchemexpress LLC Supplier No. HY133771AS10MG
Nordoxepin-d3 hydrochloride is the deuterium-labeled form of nordoxepin hydrochloride, the active metabolite of doxepin. It is provided as a solid for use as a tracer and internal standard in quantitative analytical workflows, including NMR, GC-MS, and LC-MS.
  • High chemical purity (99.9%).
  • Deuterium-labeled for accurate internal standardization.
  • Suitable for NMR, GC-MS, and LC-MS quantitation.
  • Supplied as a stable solid for convenient handling and storage.
  • Useful as a tracer in pharmacokinetic and metabolite studies.