Learn More
Medchemexpress LLC Nordoxepin-d4 hydrochloride | 2249819-14-3 | 99.9% | 305.84 | C18H16D4ClNO | 1 MG
SDP
Catalog No. 5000215684
Encompass_Preferred
Nordoxepin-d4 hydrochloride is the deuterium-labeled hydrochloride salt of nordoxepin provided for research and analytical use. It is intended primarily as an isotopically labeled internal standard or tracer in bioanalytical, pharmacokinetic, and metabolic studies, and is optimized for accurate quantification in mass spectrometry workflows.
  • High isotopic and chemical purity (99.9%).
  • Suitable as an internal standard for LC-MS/MS and GC-MS assays.
  • Useful for bioanalytical quantification and method validation.
  • Deuterium labeling enables tracer and metabolic studies.
  • Hydrochloride form improves handling and stability.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-15684 Supplier Medchemexpress LLC Supplier No. HY100046S11MG
Nordoxepin-d4 hydrochloride is the deuterium-labeled hydrochloride salt of nordoxepin provided for research and analytical use. It is intended primarily as an isotopically labeled internal standard or tracer in bioanalytical, pharmacokinetic, and metabolic studies, and is optimized for accurate quantification in mass spectrometry workflows.
  • High isotopic and chemical purity (99.9%).
  • Suitable as an internal standard for LC-MS/MS and GC-MS assays.
  • Useful for bioanalytical quantification and method validation.
  • Deuterium labeling enables tracer and metabolic studies.
  • Hydrochloride form improves handling and stability.
  • For research use only; not for human or veterinary use.