Learn More
Medchemexpress LLC Nordoxepin-d4 hydrochloride | 2249819-14-3 | 99.9% | 305.84 | C18H16D4ClNO | 10 MG
SDP
Supplier:  Medchemexpress LLC HY100046S110MG
Encompass_Preferred
Nordoxepin-d4 hydrochloride is a deuterium-labeled reference standard of nordoxepin hydrochloride for research and analytical use. It is formulated for use as an isotopically labeled internal standard to support accurate quantitation, method validation, and metabolic or pharmacokinetic studies by LC-MS.
  • High isotopic enrichment for precise internal standardization.
  • High purity (99.9%) suitable for quantitative analysis.
  • Compatible with LC-MS workflows and method development.
  • Supplied in small, analytical-scale pack sizes for convenience.
  • Provides defined molecular weight and formula for reference.
Catalog No. 50-002-15390