Learn More
Medchemexpress LLC Nordoxepin-d4 hydrochloride | 2249819-14-3 | 99.9% | 305.84 g/mol | C18H16D4ClNO | 5 MG
SDP
Catalog No. 5000215428
Encompass_Preferred
Nordoxepin-d4 hydrochloride is the deuterium-labeled analog of nordoxepin hydrochloride supplied for use as an isotopically labeled internal standard in analytical and research applications. It is provided as a high-purity solid suitable for method development, quantitation, and metabolite studies. For research use only.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity (99.9%) for reliable analytical performance.
  • Molecular formula C18H16D4ClNO; molecular weight 305.84 g/mol.
  • Available in small milligram quantities for analytical workflows.
  • Supplied as hydrochloride salt for stability in common solvents.
Catalog No. 50-002-15428 Supplier Medchemexpress LLC Supplier No. HY100046S15MG
Nordoxepin-d4 hydrochloride is the deuterium-labeled analog of nordoxepin hydrochloride supplied for use as an isotopically labeled internal standard in analytical and research applications. It is provided as a high-purity solid suitable for method development, quantitation, and metabolite studies. For research use only.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity (99.9%) for reliable analytical performance.
  • Molecular formula C18H16D4ClNO; molecular weight 305.84 g/mol.
  • Available in small milligram quantities for analytical workflows.
  • Supplied as hydrochloride salt for stability in common solvents.