Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nordoxepin hydrochloride | 2887-91-4 | 99.9% | 301.81 | C18H20ClNO | 5 MG
SDP

Catalog No. 5000215492
Encompass_Preferred

Nordoxepin hydrochloride is the major metabolite of the tricyclic antidepressant doxepin, provided as a research-grade chemical standard for analytical and laboratory applications. This compound is intended for non-clinical research use only.

  • Major metabolite of doxepin, useful as an analytical standard.
  • Purity 99.9%.
  • Molecular formula C18H20ClNO and molecular weight 301.81.
  • White to off-white solid appearance.
  • Available in small solid quantities suitable for analytical work.
  • Store sealed at 4°C; solutions are stable at -80°C for long-term storage.

Catalog No. 50-002-15492 Supplier Medchemexpress LLC Supplier No. HY1000465MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nordoxepin hydrochloride is the major metabolite of the tricyclic antidepressant doxepin, provided as a research-grade chemical standard for analytical and laboratory applications. This compound is intended for non-clinical research use only.

  • Major metabolite of doxepin, useful as an analytical standard.
  • Purity 99.9%.
  • Molecular formula C18H20ClNO and molecular weight 301.81.
  • White to off-white solid appearance.
  • Available in small solid quantities suitable for analytical work.
  • Store sealed at 4°C; solutions are stable at -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.