Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nordoxepin hydrochloride | 2887-91-4 | 99.9% | 301.81 g/mol | C18H20ClNO | 10 MG
SDP

Catalog No. 5000215306
Encompass_Preferred

Nordoxepin hydrochloride is the primary metabolite of the tricyclic antidepressant doxepin and is provided as a solid research reagent for analytical, pharmacological, and metabolic studies. It is supplied with characterized purity and storage recommendations and is intended for research use only (not for human or veterinary use).

  • Chemical identity: CAS 2887-91-4; formula C18H20ClNO; MW 301.81 g/mol.
  • High purity (99.94%).
  • Appearance: solid; store sealed and away from moisture.
  • Storage in solvent: -80°C up to 6 months; -20°C up to 1 month.
  • Intended applications: analytical and pharmacokinetic research.

Catalog No. 50-002-15306 Supplier Medchemexpress LLC Supplier No. HY10004610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nordoxepin hydrochloride is the primary metabolite of the tricyclic antidepressant doxepin and is provided as a solid research reagent for analytical, pharmacological, and metabolic studies. It is supplied with characterized purity and storage recommendations and is intended for research use only (not for human or veterinary use).

  • Chemical identity: CAS 2887-91-4; formula C18H20ClNO; MW 301.81 g/mol.
  • High purity (99.94%).
  • Appearance: solid; store sealed and away from moisture.
  • Storage in solvent: -80°C up to 6 months; -20°C up to 1 month.
  • Intended applications: analytical and pharmacokinetic research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.