Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nordoxepin hydrochloride | 2887-91-4 | 99.9% | 301.81 g/mol | C18H20ClNO | 25 MG
SDP

Catalog No. 5000215494
Encompass_Preferred

Nordoxepin hydrochloride is the major metabolite of the tricyclic antidepressant doxepin and is provided as an analytical standard for biochemical and pharmacological research. It is intended for laboratory use only and not for human consumption.

  • Used as an analytical standard and research reagent.
  • High purity (99.9%).
  • Molecular formula C18H20ClNO; molecular weight 301.81 g/mol.
  • Available in multiple pack sizes, including 25 mg and DMSO solutions.
  • Storage: sealed at 4°C; in solvent store at -80°C for long-term stability.

Catalog No. 50-002-15494 Supplier Medchemexpress LLC Supplier No. HY10004625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nordoxepin hydrochloride is the major metabolite of the tricyclic antidepressant doxepin and is provided as an analytical standard for biochemical and pharmacological research. It is intended for laboratory use only and not for human consumption.

  • Used as an analytical standard and research reagent.
  • High purity (99.9%).
  • Molecular formula C18H20ClNO; molecular weight 301.81 g/mol.
  • Available in multiple pack sizes, including 25 mg and DMSO solutions.
  • Storage: sealed at 4°C; in solvent store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.