Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nordoxepin hydrochloride | 2887-91-4 | 99.9% | 301.81 g/mol | C18H20ClNO | 50 MG
SDP

Catalog No. 5000215162
Encompass_Preferred

Nordoxepin hydrochloride is the major active metabolite of the tricyclic antidepressant doxepin, supplied as a research-grade reference standard for analytical and pharmacological studies.

  • Molecular formula: C18H20ClNO.
  • Molecular weight: 301.81 g/mol.
  • CAS number: 2887-91-4.
  • Purity: 99.9%.
  • Appearance: white to off-white solid.
  • Recommended storage: 4°C sealed; in solvent -80°C up to 6 months, -20°C up to 1 month.
  • Supplied amount: 50 mg.
  • Intended use: for research and analytical applications.

Catalog No. 50-002-15162 Supplier Medchemexpress LLC Supplier No. HY10004650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nordoxepin hydrochloride is the major active metabolite of the tricyclic antidepressant doxepin, supplied as a research-grade reference standard for analytical and pharmacological studies.

  • Molecular formula: C18H20ClNO.
  • Molecular weight: 301.81 g/mol.
  • CAS number: 2887-91-4.
  • Purity: 99.9%.
  • Appearance: white to off-white solid.
  • Recommended storage: 4°C sealed; in solvent -80°C up to 6 months, -20°C up to 1 month.
  • Supplied amount: 50 mg.
  • Intended use: for research and analytical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.