Learn More
Sigma Aldrich Fine Chemicals Biosciences NorellR natural quartz 5 m
Catalog No. 502904944
Encompass
Norell clear fuzed quartz tubes 99.8% SiO2 are recommended for Boron NMR spectroscopy (0.1 ppm Boron). Also for applications requiring temperatures above 150C and/or UV catalyzed reactions at wavelength above 255 nm (80% T 255 nm).
Catalog No. 50-290-4944 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. NORS5500QTZ75EA
Norell clear fuzed quartz tubes 99.8% SiO2 are recommended for Boron NMR spectroscopy (0.1 ppm Boron). Also for applications requiring temperatures above 150C and/or UV catalyzed reactions at wavelength above 255 nm (80% T 255 nm).