Learn More
Sigma Aldrich Fine Chemicals Biosciences NorellR Sample VaultTM NMR
Catalog No. 502901407
Encompass
Sample Vault caps have superior holding and sealing capabilities which eliminate cap/tube failure in your instrument. The patented design incorporates a white band positioned on the NMR tube that aligns with the base of the cap indicating a properly locked position.The superior Sample Vault cap attaches semi-permanently for multiple use and for critical applications.
Catalog No. 50-290-1407 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z69335996EA
Sample Vault caps have superior holding and sealing capabilities which eliminate cap/tube failure in your instrument. The patented design incorporates a white band positioned on the NMR tube that aligns with the base of the cap indicating a properly locked position.The superior Sample Vault cap attaches semi-permanently for multiple use and for critical applications.