Learn More
Sigma Aldrich Fine Chemicals Biosciences NorellR Select SeriesTM 3
Catalog No. 502901080
Encompass
The high-throughput NMR tubes are ideal for reducing salt effects when running buffered SOLUTIONons for bio-samples. They are made from ASTM Type 1 Class A borosilicate glass for reproducibility and durability and are specially designed for one time use for routine NMR.
Catalog No. 50-290-1080 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. NRS3HT725EA
The high-throughput NMR tubes are ideal for reducing salt effects when running buffered SOLUTIONons for bio-samples. They are made from ASTM Type 1 Class A borosilicate glass for reproducibility and durability and are specially designed for one time use for routine NMR.