Learn More
Sigma Aldrich Fine Chemicals Biosciences NorellR Select SeriesTM 5
Supplier:  Sigma Aldrich Fine Chemicals Biosciences NORS550075EA
Encompass
Manufactured out of ASTM Type 1 Class A glass Norell Select Series NMR tubes is specifically designed for high-reSOLUTIONon NMR spectroscopy. The Select Series provides reliable performance for spinning or non-spinning techniques structure determination of small molecules or determining the 3-D structure of a membrane protein.
Catalog No. 50-290-1479