Learn More
Sigma Aldrich Fine Chemicals Biosciences NorellR Standard SeriesTM
Supplier:  Sigma Aldrich Fine Chemicals Biosciences NOR509UP75EA
Encompass
The Norell Standard Series NMR tubes are redesigned to eliminate tube wall and bottom variations. This improves shimming times and sample reproducibility.Manufactured out of ASTM Type 1 Class B glass or commonly referred to as N-51A these NMR tubes are designed for auto-sampling systems and high-throughput NMR spectroscopy.
Catalog No. 50-290-1897