Learn More
Sigma Aldrich Fine Chemicals Biosciences NorellR valved 5 mm NMR tu1EA
Catalog No. 502904677
Encompass
These NMR tubes feature a glass/PTfE pressure valve permanently joined to a choice of available sample tubes. These pressure tubes facilitate experiments requiring conditions such as pressurized inert atmosphere blanketing addition of reactive gaseous reagents under pressure containment of low boiling point solvents or samples at elevated temperatures.
Catalog No. 50-290-4677 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. NRS5500MWIPV81EA
These NMR tubes feature a glass/PTfE pressure valve permanently joined to a choice of available sample tubes. These pressure tubes facilitate experiments requiring conditions such as pressurized inert atmosphere blanketing addition of reactive gaseous reagents under pressure containment of low boiling point solvents or samples at elevated temperatures.