Learn More
Selleck Chemical LLC (−)-Norepinephrine
SDP
Supplier:  Selleck Chemical LLC S950725MG
Encompass_Preferred
(−)-Norepinephrine (Arterenol, Levarterenol, L-Noradrenaline) is a natural neurotransmitter and hormone. It is an agonist of adrenergic receptors with Ki values of 330, 56, and 740 nM for α1, α2, and β1 adrenoceptors, respectively.
Catalog No. 50-238-9711