Learn More
Medchemexpress LLC Norepinephrine | 51-41-2 | 100.0% | 169.18 | 100 MG
SDP
Catalog No. 5000351093
Encompass_Preferred
Norepinephrine (Standard) is the analytical standard for Norepinephrine, intended for research and analytical applications. It is a potent adrenergic receptor (AR) agonist that activates α1, α2, and β1 receptors.
  • Analytical standard for Norepinephrine
  • Potent adrenergic receptor agonist
  • Activates α1, α2, and β1 receptors
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-003-51093 Supplier Medchemexpress LLC Supplier No. HY13715R100MG
Norepinephrine (Standard) is the analytical standard for Norepinephrine, intended for research and analytical applications. It is a potent adrenergic receptor (AR) agonist that activates α1, α2, and β1 receptors.
  • Analytical standard for Norepinephrine
  • Potent adrenergic receptor agonist
  • Activates α1, α2, and β1 receptors
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications