Learn More
Medchemexpress LLC Norepinephrine (hydrochloride) | 329-56-6 | 99.95% | 205.64 | 1 ML
SDP
Catalog No. 5000348840
Encompass_Preferred
Norepinephrine hydrochloride is a potent adrenergic receptor (AR) agonist that activates α1, α2, and β1 receptors. It is generally considered a β1-subtype selective adrenergic agonist, but also shows direct activity at the β2-adrenoceptor at higher concentrations. This product is suitable for various laboratory applications, including studies on cAMP increase and lipolysis in adipocytes. It can also be used to induce cardiomyopathy models in male Sprague-Dawley rats via continuous injection.
  • Activates α1, α2, and β1 adrenergic receptors
  • Increases cAMP and induces lipolysis in adipocytes
  • Suitable for cardiomyopathy induction models
  • For research use only
Catalog No. 50-003-48840 Supplier Medchemexpress LLC Supplier No. HY13715A10MM/1ML
Norepinephrine hydrochloride is a potent adrenergic receptor (AR) agonist that activates α1, α2, and β1 receptors. It is generally considered a β1-subtype selective adrenergic agonist, but also shows direct activity at the β2-adrenoceptor at higher concentrations. This product is suitable for various laboratory applications, including studies on cAMP increase and lipolysis in adipocytes. It can also be used to induce cardiomyopathy models in male Sprague-Dawley rats via continuous injection.
  • Activates α1, α2, and β1 adrenergic receptors
  • Increases cAMP and induces lipolysis in adipocytes
  • Suitable for cardiomyopathy induction models
  • For research use only