Learn More
Medchemexpress LLC Norepinephrine (Standard) | 51-41-2 | 100.0% | 169.18 | 50 MG
SDP
Catalog No. 5000352063
Encompass_Preferred
Norepinephrine (Standard) is the analytical standard of Norepinephrine. It is intended for research and analytical applications. It is a potent adrenergic receptor (AR) agonist, activating α1, α2, and β1 receptors. The compound is the grade of analytical standard, which is the reference standard supplied for assay. It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for norepinephrine, with a high purity of 99.97%.
  • Intended exclusively for research and analytical applications.
  • Utilized in qualitative, quantitative, and methodological research experiments, particularly with techniques such as high-performance liquid chromatography (HPLC), gas chromatography (GC), and mass spectrometry (MS).
  • Not for sale to patients.
Catalog No. 50-003-52063 Supplier Medchemexpress LLC Supplier No. HY13715R50MG
Norepinephrine (Standard) is the analytical standard of Norepinephrine. It is intended for research and analytical applications. It is a potent adrenergic receptor (AR) agonist, activating α1, α2, and β1 receptors. The compound is the grade of analytical standard, which is the reference standard supplied for assay. It is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for norepinephrine, with a high purity of 99.97%.
  • Intended exclusively for research and analytical applications.
  • Utilized in qualitative, quantitative, and methodological research experiments, particularly with techniques such as high-performance liquid chromatography (HPLC), gas chromatography (GC), and mass spectrometry (MS).
  • Not for sale to patients.