Learn More
Medchemexpress LLC Norepinephrine (tartrate) | 51-40-1 | 100.0% | 319.26 | 100 MG
SDP
Catalog No. 5000350718
Encompass_Preferred
Norepinephrine (Levarterenol; L-Noradrenaline) tartrate is a potent adrenergic receptor (AR) agonist that activates α1, α2, and β1 receptors. It is intended for research use only. This compound is known to induce cardiomyopathy in animal models and plays a role in cellular processes such as increasing cAMP and inducing lipolysis in adipocytes.
  • Potent adrenergic receptor agonist
  • Activates α1, α2, and β1 receptors
  • Induces cardiomyopathy in animal models
  • Increases cAMP and induces lipolysis in adipocytes
Catalog No. 50-003-50718 Supplier Medchemexpress LLC Supplier No. HY13715C100MG
Norepinephrine (Levarterenol; L-Noradrenaline) tartrate is a potent adrenergic receptor (AR) agonist that activates α1, α2, and β1 receptors. It is intended for research use only. This compound is known to induce cardiomyopathy in animal models and plays a role in cellular processes such as increasing cAMP and inducing lipolysis in adipocytes.
  • Potent adrenergic receptor agonist
  • Activates α1, α2, and β1 receptors
  • Induces cardiomyopathy in animal models
  • Increases cAMP and induces lipolysis in adipocytes