Learn More
Medchemexpress LLC Norepinephrine tartrate | 51-40-1 | 99.96% | 319.26 | 500 MG
SDP
Catalog No. 5000349019
Encompass_Preferred
Norepinephrine (Levarterenol; L-Noradrenaline) tartrate is a potent adrenergic receptor (AR) agonist that activates α1, α2, and β1 receptors. It is generally considered a β1-subtype selective adrenergic agonist over β2-adrenoceptor, but also has direct activity at the β2-adrenoceptor in higher concentrations.
  • Potent adrenergic receptor (AR) agonist
  • β1-subtype selective
  • For research purposes only
  • High purity: 99.96%
  • Used in various research areas
  • Can be used to induce cardiomyopathy in animal models
  • Involved in multiple clinical trials for various conditions
Catalog No. 50-003-49019 Supplier Medchemexpress LLC Supplier No. HY13715C500MG
Norepinephrine (Levarterenol; L-Noradrenaline) tartrate is a potent adrenergic receptor (AR) agonist that activates α1, α2, and β1 receptors. It is generally considered a β1-subtype selective adrenergic agonist over β2-adrenoceptor, but also has direct activity at the β2-adrenoceptor in higher concentrations.
  • Potent adrenergic receptor (AR) agonist
  • β1-subtype selective
  • For research purposes only
  • High purity: 99.96%
  • Used in various research areas
  • Can be used to induce cardiomyopathy in animal models
  • Involved in multiple clinical trials for various conditions