Learn More
Cayman Chemical (±)-Norepinephrine-d6 (hydrochloride), Purity: ≥99% (deuterated forms d1-d6), CAS: 1219803-04-9, 10 mg
SDP
Catalog No. 502951104
Encompass_Preferred
An internal standard for the quantification of norepinephrine by GC- or LC-MS
Catalog No. 50-295-1104 Supplier Cayman Chemical Supplier No. 1084210 MG
An internal standard for the quantification of norepinephrine by GC- or LC-MS