Learn More
Cayman Chemical (–)-Norepinephrine (bitartrate hydrate), Purity: ≥95%, CAS: 108341-18-0, 100 mg
SDP
Catalog No. 502968449
Encompass_Preferred
A natural neurotransmitter and hormone that acts as an agonist of adrenergic receptors (Ki values are 330, 56, and 740 nM for α1, α2, and β1 adrenoceptors, respectively)
Catalog No. 50-296-8449 Supplier Cayman Chemical Supplier No. 16673100 MG
A natural neurotransmitter and hormone that acts as an agonist of adrenergic receptors (Ki values are 330, 56, and 740 nM for α1, α2, and β1 adrenoceptors, respectively)